missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Chloro-4-(trifluoromethyl)benzaldehyde is used in synthesis of organic compunds in Îł-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
3-Chloro-4-(trifluoromethyl)benzaldehyde is used in synthesis of organic compunds in Îł-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Specifications
Specifications
Refractive Index | 1.4944 |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 208.57 |
Percent Purity | 97% |
Chemical Name or Material | 3-Chloro-4-(trifluoromethyl)benzaldehyde |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10158886 | Certificate of Analysis | 08/12/2021 | H31635-03,H31635.06 |
10202947 | Certificate of Analysis | 08/12/2021 | H31635-03,H31635.06 |
10153633 | Certificate of Analysis | 06/12/2021 | H31635-03,H31635.06 |
10167645 | Certificate of Analysis | 06/12/2021 | H31635-03,H31635.06 |
10172924 | Certificate of Analysis | 06/12/2021 | H31635-03,H31635.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction