missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™ Product Code.: 15454237

3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15454237
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15454237

Brand: Thermo Scientific Alfa Aesar H31635.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-4-(trifluoromethyl)benzaldehyde is used in synthesis of organic compunds in γ-Aminobutyrate-A Receptor Modulation by 3-Aryl-1-(arylsulfonyl)- 1,4,5,6-tetrahydropyridazines.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83279-38-3
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD03095197
InChI Key UTSBZTCYCRVHCK-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl benzaldehyde, benzaldehyde, 3-chloro-4-trifluoromethyl, benzaldehyde,3-chloro-4-trifluoromethyl, 3-chloro-4-trifluoromethyl benzaldehyde, jrd
PubChem CID 19031287
IUPAC Name 3-chloro-4-(trifluoromethyl)benzaldehyde
SMILES FC(F)(F)C1=C(Cl)C=C(C=O)C=C1

Specifications

Refractive Index 1.4944
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 208.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-4-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.