missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline hydrochloride, 97%, Thermo Scientific™

Product Code. 11383097
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-85-5
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00035485
InChI Key NMGHWHCTRGZZOP-UHFFFAOYSA-N
Synonym 3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
PubChem CID 67336
IUPAC Name 3-chloroaniline;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)N.Cl

Specifications

Melting Point ∽222°C (decomposition)
Odor Characteristic
Quantity 25 g
UN Number UN2018
Beilstein 3623168
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 3-Chloroaniline hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'