missing translation for 'onlineSavingsMsg'
Learn More
3-Fluoro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™ Product Code.: 11440787

3-Fluoro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11440787
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11440787

Brand: Thermo Scientific Alfa Aesar B23189.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213203-65-7
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.03
MDL Number MFCD00236289
InChI Key MOVSRIIBGRAUAF-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzyl bromide, 4-bromomethyl-2-fluoro-1-trifluoromethyl benzene, 4-bromomethyl-2-fluoro-1-trifluoromethyl-benzene, acmc-20ai27, 3-fluoro-4-trifluoromethylbenzyl bromide, 3-fluoro-4-trifluoromethyl benzylbromide, 3-fluoro-4-trifluoromethyl-benzyl bromide, benzene, 4-bromomethyl-2-fluoro-1-trifluoromethyl, benzene,4-bromomethyl-2-fluoro-1-trifluoromethylShow MoreShow Less
PubChem CID 2737574
IUPAC Name 4-(bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene
SMILES FC1=CC(CBr)=CC=C1C(F)(F)F

Specifications

Melting Point 25°C to 27°C
Quantity 1 g
UN Number UN1760
Sensitivity Moisture sensitive
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzyl bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10182796 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number61200411 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10164281 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.