Learn More
3-Fluorobenzonitrile, 98%, Thermo Scientific Chemicals Product Code.: 11475397
3-Fluorobenzonitrile, 98%, Thermo Scientific Chemicals
Product Code. 11475397
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
Product Code. 11475397
Brand: Thermo Scientific Alfa Aesar B23426.06
Request Bulk or Custom Format
CAS: 403-54-3 | C7H4FN | 121.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 403-54-3
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001797
InChI Key JZTPKAROPNTQQV-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitileShow MoreShow Less
PubChem CID 67880
IUPAC Name 3-fluorobenzonitrile
SMILES FC1=CC=CC(=C1)C#N
Specifications
Melting Point -16°C
Density 1.14
Boiling Point 183°C
Flash Point 67°C (152°F)
Odor Almond-like
Refractive Index 1.5055
Quantity 5 g
UN Number UN3276
Beilstein 1858941
Formula Weight 121.11
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzonitrile
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254894 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberB23426.22,B23426.06,B23426.14
Lot Number10205036 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberB23426.22,B23426.06,B23426.14
Lot Number10178586 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23426.22,B23426.06,B23426.14
Lot NumberJ26Q040 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23426.22,B23426.06,B23426.14
Lot Number10100326 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23426.22,B23426.06,B23426.14
Product Title
3-Fluorobenzonitrile, 98%, Thermo Scientific Chemicals >