Learn More
3'-Methylbiphenyl-3-carboxylic acid, 97%, Thermo Scientific™
33.35€ - 130.00€
Chemical Identifiers
CAS | 158619-46-6 |
---|---|
Molecular Formula | C14H11O2 |
Molecular Weight (g/mol) | 211.24 |
MDL Number | MFCD00558892 |
InChI Key | BJVLFJADOVATRU-UHFFFAOYSA-M |
Synonym | 3'-methylbiphenyl-3-carboxylic acid, 3-3-methylphenyl benzoic acid, 3'-methyl 1,1'-biphenyl-3-carboxylic acid, 3'-methyl-1,1'-biphenyl-3-carboxylic acid, 3'-methyl-biphenyl-3-carboxylic acid, acmc-1ccsu, 3-m-tolyl benzoic acid, amtda074, cambridge id 5211690, 3-3-carboxyphenyl toluene |
PubChem CID | 747832 |
IUPAC Name | 3-(3-methylphenyl)benzoic acid |
SMILES | CC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
158619-46-6 | |
211.24 | |
BJVLFJADOVATRU-UHFFFAOYSA-M | |
747832 | |
CC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O |
C14H11O2 | |
MFCD00558892 | |
3'-methylbiphenyl-3-carboxylic acid, 3-3-methylphenyl benzoic acid, 3'-methyl 1,1'-biphenyl-3-carboxylic acid, 3'-methyl-1,1'-biphenyl-3-carboxylic acid, 3'-methyl-biphenyl-3-carboxylic acid, acmc-1ccsu, 3-m-tolyl benzoic acid, amtda074, cambridge id 5211690, 3-3-carboxyphenyl toluene | |
3-(3-methylphenyl)benzoic acid |
Specifications
158619-46-6 | |
MFCD00558892 | |
Slightly soluble in water. | |
CC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O | |
211.24 | |
212.25 | |
3'-Methylbiphenyl-3-carboxylic acid |
C14H11O2 | |
3'-methylbiphenyl-3-carboxylic acid, 3-3-methylphenyl benzoic acid, 3'-methyl 1,1'-biphenyl-3-carboxylic acid, 3'-methyl-1,1'-biphenyl-3-carboxylic acid, 3'-methyl-biphenyl-3-carboxylic acid, acmc-1ccsu, 3-m-tolyl benzoic acid, amtda074, cambridge id 5211690, 3-3-carboxyphenyl toluene | |
BJVLFJADOVATRU-UHFFFAOYSA-M | |
3-(3-methylphenyl)benzoic acid | |
747832 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.