missing translation for 'onlineSavingsMsg'
Learn More
3-Methylthiophene-2-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11439476

3-Methylthiophene-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11439476
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11439476

Brand: Thermo Scientific Alfa Aesar A19194.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23806-24-8
Molecular Formula C6H5O2S
Molecular Weight (g/mol) 141.16
MDL Number MFCD00005438
InChI Key IFLKEBSJTZGCJG-UHFFFAOYSA-M
Synonym 3-methyl-2-thiophenecarboxylic acid, 3-methyl-2-thenoic acid, 2-thiophenecarboxylic acid, 3-methyl, 2-carboxy-3-methylthiophene, 3-methyl-2-thiophene carboxylic acid, 3-methyl-2-thiophenecarboxylic, 3-methyl-thiophene-2-carboxylic acid, 3-methyl-2-thiophenecarboxylicacid, pubchem5182, acmc-1cinc
PubChem CID 90269
IUPAC Name 3-methylthiophene-2-carboxylic acid
SMILES CC1=C(SC=C1)C([O-])=O

Specifications

Melting Point 144°C to 149°C
Quantity 25 g
Beilstein 116184
Formula Weight 142.18
Percent Purity 98%
Chemical Name or Material 3-Methylthiophene-2-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10147036 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19194.06,A19194.14,A19194.22
Lot Number10106334 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19194.06,A19194.14,A19194.22
Lot NumberT17E021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19194.06,A19194.14,A19194.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methylthiophene-2-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.