Learn More
3-Nitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Description
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.
Solubility
Hydrolyzes slowly in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.
Specifications
Specifications
Melting Point | 59°C to 63°C |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
UN Number | UN3261 |
Beilstein | 746542 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes slowly in water. |
Formula Weight | 221.62 |
Percent Purity | 98% |
Chemical Name or Material | 3-Nitrobenzenesulfonyl chloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.