missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)benzylamine, 97%, Thermo Scientific™

Product Code. 15407447
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771573-34-3
Molecular Formula C7H8F5NS
Molecular Weight (g/mol) 233.2
MDL Number MFCD06213674
InChI Key FJOVSWPDVAFUMX-UHFFFAOYSA-N
Synonym 3-pentafluorosulfur benzylamine, 1-3-pentafluoro-??-sulfanyl phenyl methanamine, 3-pentafluorothio benzylamine, 3-pentafluoro-lambda-sulfanyl phenyl methanamine
PubChem CID 66523498
IUPAC Name [3-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)S(F)(F)(F)(F)F)CN

Specifications

Quantity 5 g
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 233.2
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'