missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals

Product Code. 11332476
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1504-58-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00040914
InChI Key NITUNGCLDSFVDL-UHFFFAOYSA-N
Synonym 3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
PubChem CID 123115
IUPAC Name 3-phenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CCO

Specifications

Density 1.078
Boiling Point 129°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5867
Quantity 5 g
Beilstein 2040911
Formula Weight 132.16
Percent Purity 97%
Chemical Name or Material 3-Phenyl-2-propyn-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Phenyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.