missing translation for 'onlineSavingsMsg'
Learn More

3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals

40.40€ - 405.02€

Chemical Identifiers

CAS 6480-68-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00006770
InChI Key DJXNJVFEFSWHLY-UHFFFAOYSA-N
Synonym 3-quinolinecarboxylic acid, 3-quinoline carboxylic acid, 3-quinolinecarboxylicacid, pubchem7517, 3-quinolylcarboxylic acid, acmc-1bfc6, 3-quinolinecarboxylic acid 1g, 3-quinolinecarboxylic acid, purum hplc, q3cShow MoreShow Less
PubChem CID 80971
IUPAC Name quinoline-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C=N2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10682095
SDS View Documents
Thermo Scientific Acros
175700010
1 g Glass bottle
40.40€
1g
Please to purchase this item. Need a web account? Register with us today!
10194330
SDS View Documents
Thermo Scientific Acros
175700050
5 g Glass bottle
122.61€
5g
Please to purchase this item. Need a web account? Register with us today!
10701945
SDS View Documents
Thermo Scientific Acros
175700250
25 g Glass bottle
405.02€
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.