Learn More
Hamilton™ 30 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Brand:  Hamilton™ 22030-01
Product Code. 12381818
  • 145.00€ / Pack of 6
Estimated Shipment: 23-12-2024
Description
Specification Sheet
Gauge30 gauge
Point Style2, 3, or 4
Needle LengthCustom Needle Length
AutoclavableYes
Sales Quantity6 needles per pack
HubNo Hub
Specifications
Specifications
30 G
Hubless Needles