Learn More
TubeWriter 300 - Custom Rack Top Print for Any Labware
Custom racks made to exact specifications to print directly on the top of unique labware
Brand:  TubeWriter PT-0000XX-300-Custom
Product Code. 30067441
  • 3090.00€ / Each
Estimated Shipment: 24-01-2025
Description
  • Custom racks designed for 300
  • Racks made to your exact specifications to directly on the top of any labware
  • Rack will ensure correct alignment and holds labware securely
  • No time-consuming sticky labels or handwriting required
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Specifications
Labeling Rack