Learn More
3M™ Versaflo™ Length-Adjusting Breathing Tube
Features a flexible helix that adjusts to the appropriate length for the individual wearer.
Brand: 3M™ 7100006955
64.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- QRS (Quick Release Swivel) breathing tubes allow fast and easy connection
- Meets higher respiratory performance requirements with 3M™ headtops
- Special flame-resistant sleeve lengthens life of tube and protects it from flying sparks
- Disposable breathing tube cover is available to help cleaning and decontamination operations
- 3M Catalog# BT-30
Specifications
Breathing Tube | |
Assembly, facility maintenance, grinding, machine operation, painting, sanding | |
Versaflo™ TR-300 Series PAPR | |
BT-30 |
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
3M™ Versaflo™ Length-Adjusting Breathing Tube > Length Adjusting Breathing Tube