Learn More
3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Outer shrouds are replacement parts for use with M-400 helmets.
Brand:  3M™ 7100137905
97.68 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15790068
  • 152.00€ / Each
Only null left
Estimated Shipment: 02-07-2025
Description
  • The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
  • 3M Catalog# M-445, M-446, M-447, M-448
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1,000
Outer Shroud
M-400 helmet
High Durability
Cordura Nylon
Cleaning, electrical, facility maintenance, painting, sanding, welding
M-448
Videos
  • 3M - Powered and supplied air products
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M Powered and supplied air products
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    3M™ Versaflo™ M-400 Series PAPR Outer Shroud > Style: High Durability