missing translation for 'onlineSavingsMsg'
Learn More

Enzymaster (3S,6S)-3,6-Octanediol, >99%

(3S,6S)-3,6-Octanediol with very high chemical and optical purities >99%.

Brand:  Enzymaster 1052-5

 View more versions of this product

Product Code. 17894651

  • 999.00€ / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Enzymatically produced
  • Chemical purity >99%
  • Enantiomeric excess >99%
  • Diastereomeric excess >99%
Specifications

Specifications

49°C
>99%
146.23 g/mol
Reagent
>99%
Off-white
5 g
>99%
Crystalline
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Enzymaster (3S,6S)-3,6-Octanediol, >99% > (3S,6S)-3,6-Octanediol, >99%; Quantity: 5 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.