missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Tetrazol-5-yl)benzyl alcohol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Quantity | 1 g |
Formula Weight | 176.18 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-Tetrazol-5-yl)benzyl alcohol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10153884 | Certificate of Analysis | 06/12/2021 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(1H-Tetrazol-5-yl)benzyl alcohol, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction