missing translation for 'onlineSavingsMsg'
Learn More

4-(2,6-Dimethylphenyl)-3-thiosemicarbazide, 97%, Thermo Scientific™

Product Code. 11358685
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71058-35-0
Molecular Formula C9H13N3S
Molecular Weight (g/mol) 195.28
MDL Number MFCD00041281
InChI Key YEOAXWBYZFHOIZ-UHFFFAOYSA-N
Synonym 4-2,6-dimethylphenyl-3-thiosemicarbazide, 3-amino-1-2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl hydrazinecarbothioamide, 1-amino-3-2,6-dimethylphenyl thiourea, 2,6-dimethylphenylthiosemicarbazide, 2,6-dimethylphenyl amino hydrazinomethane-1-thione
PubChem CID 736213
IUPAC Name 3-amino-1-(2,6-dimethylphenyl)thiourea
SMILES CC1=CC=CC(C)=C1NC(=S)NN

Specifications

Melting Point ∽225°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 6768435
Formula Weight 195.29
Percent Purity 97%
Chemical Name or Material 4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'