Learn More
Memmert™ 4 - 20 mA Current Loop Interface
4-20mA current loop interface temperature controller
Brand:  Memmert™ IPP2200ecoV3
 View more versions of this product
Product Code. 17120165
  • 242.00€ / Each
Estimated Shipment: 08-08-2025
Description
  • Temperature controller
  • Actual value; -10 to +80 °C = 4 -20 mA
Specifications
Specifications
IPP2200eco
Temperature Controller
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ 4 - 20 mA Current Loop Interface > IPP2200eco