Learn More
Memmert™ 4-20 mA Current Loop Interface for UF55plus
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF55plusV6
Product Code. 17519525
  • 451.00€ / Each
Only null left
Estimated Shipment: 09-07-2025
Add to basket
Description
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UF55plus Oven
Current Loop Interface
4 to 20 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF55plus > For Use With: UF55plus