missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals

Product Code. 11313416
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

4,4'-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13676-54-5
Molecular Formula C21H14N2O4
Molecular Weight (g/mol) 358.353
MDL Number MFCD00005507
InChI Key XQUPVDVFXZDTLT-UHFFFAOYSA-N
Synonym bismaleimide, bismaleimide s, bis 4-maleimidophenyl methane, diphenylmethanebismaleimide, p,p'-dimaleimidodiphenylmethane, 4,4'-methylenebis n-phenylmaleimide, bis p-maleimidophenyl methane, 4,4'-dimaleimidodiphenylmethane, 4,4'-diphenylmethanedimaleimide, 4,4'-biphenylmethanebismaleimide
PubChem CID 83648
IUPAC Name 1-[4-[[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl]phenyl]pyrrole-2,5-dione
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N3C(=O)C=CC3=O)N4C(=O)C=CC4=O

Specifications

Melting Point 156°C to 158°C
Quantity 5 g
UN Number UN2811
Beilstein 333986
Formula Weight 358.36
Percent Purity 95%
Chemical Name or Material 4,4'-Methylenebis(N-phenylmaleimide)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'