missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methyl-6-nitroaniline, 97%

Product Code. 11446867
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 77811-44-0 | C7H7BrN2O2 | 231.049 g/mol

It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

Solubility
Slightly soluble in water and soluble in hot methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77811-44-0
Molecular Formula C7H7BrN2O2
Molecular Weight (g/mol) 231.049
MDL Number MFCD00052919
InChI Key ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
Synonym 4-bromo-2-methyl-6-nitro-phenylamine, 4-bromo-2-methyl-6-nitrobenzenamine, 2-amino-5-bromo-3-nitrotoluene, 4-bromo-6-nitro-o-toluidine, benzenamine, 4-bromo-2-methyl-6-nitro, 4-bromo-2-methyl-6-nitrophenylamine, 4-bromo-2-methyl-6-nitro-aniline, pubchem3813, acmc-1bg8u, ksc496e7t
PubChem CID 522679
IUPAC Name 4-bromo-2-methyl-6-nitroaniline
SMILES CC1=CC(=CC(=C1N)[N+](=O)[O-])Br

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
Beilstein 3530524
Solubility Information Slightly soluble in water and soluble in hot methanol.
Formula Weight 231.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methyl-6-nitroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-2-methyl-6-nitroaniline, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.