missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals Product Code.: 11328215

4-Chloro-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11328215
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11328215

Brand: Thermo Scientific Alfa Aesar L08533.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7697-29-2 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-29-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045853
InChI Key MRUKIIWRMSYKML-UHFFFAOYSA-N
Synonym 4-chloro-m-toluic acid, 4-chloro-3-methyl-benzoic acid, 4-chloro-3-methyl benzoic acid, benzoic acid, 4-chloro-3-methyl, m-toluic acid, 4-chloro, pubchem3666, 5-carboxy-2-chlorotoluene, acmc-1bg77, ksc497q5r, 4-chloranyl-3-methyl-benzoic acid
PubChem CID 282989
IUPAC Name 4-chloro-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Cl)C(O)=O

Specifications

Melting Point 212°C to 216°C
Quantity 2 g
Beilstein 1936535
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylbenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10124262 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL08533.18,L08533.09,L08533.04
Lot NumberGBFA018483 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL08533.18,L08533.09,L08533.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.