missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Product Code. 15456627
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108831-68-1
Molecular Formula C8H7ClN2S
Molecular Weight (g/mol) 198.67
MDL Number MFCD00464832
InChI Key HYOBKTVPACQCBR-UHFFFAOYSA-N
Synonym 4-chloro-5,6-dimethylthieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl-thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5,6-dimethyl, thieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl, 4-chloro-5,6-dimethylthiopheno 2,3-d pyrimidine, acmc-209vig, 4-chloranyl-5,6-dimethyl-thieno 2,3-d pyrimidine
PubChem CID 290212
IUPAC Name 4-chloro-5,6-dimethylthieno[2,3-d]pyrimidine
SMILES CC1=C(C)C2=C(Cl)N=CN=C2S1

Specifications

Melting Point 109°C to 110°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 198.68
Percent Purity 96%
Chemical Name or Material 4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.