missing translation for 'onlineSavingsMsg'
Learn More
4-Chlorobenzoic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11442108

4-Chlorobenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11442108
1000 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11442108

Brand: Thermo Scientific Alfa Aesar A15135.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

Solubility
Soluble in water(0.08g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-11-3
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002531
InChI Key XRHGYUZYPHTUJZ-UHFFFAOYSA-N
Synonym p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italianShow MoreShow Less
PubChem CID 6318
ChEBI CHEBI:30747
IUPAC Name 4-chlorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)Cl

Specifications

Melting Point 239°C to 243°C
Density 1.54
Boiling Point 275°C
Flash Point 238°C (460°F)
Odor Odorless
Quantity 1000 g
Beilstein 907196
Merck Index 14,2125
Solubility Information Soluble in water(0.08g/L).
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzoic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10210013 Certificate TypeCertificate of Analysis Date27/07/2023 Catalog NumberA15135.0I,A15135.30,A15135.0B
Lot Number10245670 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberA15135.30,A15135.0B
Lot Number10152045 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15135.30,A15135.0B
Lot Number10196389 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15135.30,A15135.0B
Lot Number10182753 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15135.30,A15135.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chlorobenzoic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.