missing translation for 'onlineSavingsMsg'
Learn More
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals Product Code.: 10366410

4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10366410
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10366410

Brand: Thermo Scientific 110090500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-67-2
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.59
MDL Number MFCD00000632
InChI Key KTZVZZJJVJQZHV-UHFFFAOYSA-N
Synonym 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyreneShow MoreShow Less
PubChem CID 14085
IUPAC Name 1-chloro-4-ethenylbenzene
SMILES ClC1=CC=C(C=C)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -16°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 192°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4Cl
Refractive Index 1.5645 to 1.5665
Quantity 50 g
Beilstein 05, II, 367
Specific Gravity 1.1
Stabilizer 0.1% tert.-butylcatechol
Formula Weight 138.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chlorostyrene, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398679 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number110090100,110090500
Lot NumberA0405706 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number110090100,110090500
Lot NumberA0410159 Certificate TypeCertificate of Analysis Date06/01/2022 Catalog Number110090100,110090500
Lot NumberA0422978 Certificate TypeCertificate of Analysis Date28/09/2020 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.