Learn More
(+/-)-4-Chlorostyrene oxide, 97+%, Thermo Scientific Chemicals
Product Code. 11465910
Click to view available options
Quantity:
5 g
25 g
(±)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2788-86-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00956179
InChI Key IBWLXNDOMYKTAD-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl oxirane, 4-chlorostyrene oxide, p-chlorostyrene oxide, oxirane, 4-chlorophenyl, p-chlorophenyl oxirane, 2-4-chloro-phenyl-oxirane, +/--4-chlorostyrene oxide, benzene, 1-chloro-4-epoxyethyl, 4-chlorophenyl oxirane, 4-chloro styrene oxide
PubChem CID 197010
IUPAC Name 2-(4-chlorophenyl)oxirane
SMILES C1C(O1)C2=CC=C(C=C2)Cl
Specifications
Density 1.283
Boiling Point 230°C
Quantity 5 g
Refractive Index 1.552
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 154.59
Percent Purity 98%
Chemical Name or Material (+/-)-4-Chlorostyrene oxide
compliance-icons
Product Identifier
  • (+/-)-4-Chlorostyrene oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-(4-Chlorophenyl)oxirane