Learn More
4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
33.57€ - 415.07€
Chemical Identifiers
CAS | 100-10-7 |
---|---|
Molecular Formula | C9H11NO |
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00003381 |
InChI Key | BGNGWHSBYQYVRX-UHFFFAOYSA-N |
Synonym | 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal |
PubChem CID | 7479 |
IUPAC Name | 4-(dimethylamino)benzaldehyde |
SMILES | CN(C)C1=CC=C(C=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489223
|
Thermo Scientific Alfa Aesar
A11712.22 |
100 g |
33.57€
100g |
In Stock
Log in to see stock. |
|||||
11499223
|
Thermo Scientific Alfa Aesar
A11712.36 |
500 g |
120.60€
500g |
In Stock
Log in to see stock. |
|||||
11409233
|
Thermo Scientific Alfa Aesar
A11712.0E |
2500 g |
415.07€
2500g |
In Stock
Log in to see stock. |
|||||
Description
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.
Solubility
Soluble in alcohol, ether, chloroform, acetic acid. Insoluble in water.
Notes
Light sensitive. Stable at room temperature.
Chemical Identifiers
100-10-7 | |
149.19 | |
BGNGWHSBYQYVRX-UHFFFAOYSA-N | |
7479 | |
CN(C)C1=CC=C(C=O)C=C1 |
C9H11NO | |
MFCD00003381 | |
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal | |
4-(dimethylamino)benzaldehyde |
Specifications
100-10-7 | |
176°C to 177°C (17 mmHg) | |
Strong | |
MFCD00003381 | |
Air and light sensitive | |
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal | |
BGNGWHSBYQYVRX-UHFFFAOYSA-N | |
4-(dimethylamino)benzaldehyde | |
7479 | |
98% |
73°C to 75°C | |
164°C (327°F) | |
C9H11NO | |
606802 | |
14,3230 | |
Soluble in alcohol,ether,chloroform,acetic acid. Insoluble in water. | |
CN(C)C1=CC=C(C=O)C=C1 | |
149.19 | |
149.19 | |
4-Dimethylaminobenzaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 202-819-0
missing translation for 'rtecsNumber' : CU5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- 4-Dimethylaminobenzaldehyde
Supplemental information
- MIXTURE LIST-Contains : 4-Dimethylaminobenzaldehyde
RUO – Research Use Only