missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-methoxyphenylacetic acid, 97%

Product Code. p-7029002
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
This item is not returnable. View return policy
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886498-61-9
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.166
MDL Number MFCD06660193
InChI Key TUHNRCPWPMJXQN-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenylacetic acid, 2-4-fluoro-2-methoxyphenyl acetic acid, 4-fluoro-2-methoxy phenylacetic acid, 4-fluoro-2-methoxybenzeneacetic acid, 4-fluoro-2-methoxyphenyl acetic acid
PubChem CID 17750773
IUPAC Name 2-(4-fluoro-2-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)F)CC(=O)O

Specifications

Melting Point 106°C to 110°C
Quantity 250 mg
Formula Weight 184.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxyphenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 4-Fluoro-2-methoxyphenylacetic acid, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.