missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-(trifluoromethyl)propiophenone, 98%, Thermo Scientific™

37.97€ - 134.00€

Chemical Identifiers

CAS 239107-27-8
Molecular Formula C10H8F4O
Molecular Weight (g/mol) 220.167
MDL Number MFCD00061264
InChI Key DEKWUKRLNDKROW-UHFFFAOYSA-N
Synonym 1-4-fluoro-3-trifluoromethyl phenyl propan-1-one, 4'-fluoro-3'-trifluoromethyl propiophenone, 4-fluoro-3-trifluoromethyl propiophenone, acmc-20amic, 4-fluoro-3-3 trifluoromethyl propiophenine, 4-fluoro-3-3 trifluoromethyl propiophenone, 1-4-fluoro-3-trifluoromethyl phenyl-1-propanone, 1-propanone,1-4-fluoro-3-trifluoromethyl phenyl, 1-4-fluoranyl-3-trifluoromethyl phenyl propan-1-one
PubChem CID 2737617
IUPAC Name 1-[4-fluoro-3-(trifluoromethyl)phenyl]propan-1-one
SMILES CCC(=O)C1=CC(=C(C=C1)F)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11489147
SDS View Documents
Thermo Scientific Alfa Aesar
B20685.03
1 g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11499147
SDS View Documents
Thermo Scientific Alfa Aesar
B20685.06
5 g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Fluoro-3'-(trifluoromethyl)propiophenone, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.