missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-hydroxy-3-iodo-5-methoxybenzaldehyde, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | IC6H2(OCH3)(OH)CHO |
Quantity | 50 g |
Formula Weight | 278.04 |
Percent Purity | 97% |
Chemical Name or Material | 5-Iodovanillin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-hydroxy-3-iodo-5-methoxybenzaldehyde, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction