missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxy-3-methyl-2-butanone, tech 85% Product Code.: 15493817

4-Hydroxy-3-methyl-2-butanone, tech 85%

Product Code. 15493817
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15493817

Brand: Thermo Scientific Alfa Aesar H28483.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 3393-64-4 | C5H10O2 | 102.133 g/mol

Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo vinylation and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo “vinylation” and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3393-64-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004739
InChI Key VVSRECWZBBJOTG-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl, 3-hydroxymethyl-2-butanone, 2-butanone, 4-hydroxy-3-methyl-, +, acmc-1cngd, 3-methyl-4-hydroxy-2-butanone, vvsrecwzbbjotg-uhfffaoysa, 2-butanone,4-hydroxy-3-methyl, 4-hydroxy-3-methyl-2-butanone, technical grade
PubChem CID 18829
IUPAC Name 4-hydroxy-3-methylbutan-2-one
SMILES CC(CO)C(=O)C

Specifications

Refractive Index 1.4325
Quantity 250 mg
Formula Weight 102.13
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-Hydroxy-3-methyl-2-butanone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10136935 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28483.MD,H28483.06,H28483.03
Lot Number10195420 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28483.MD,H28483.06,H28483.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxy-3-methyl-2-butanone, tech 85% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.