missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrobenzo-15-crown-5, 99%, Thermo Scientific Chemicals Product Code.: 11413337

4-Nitrobenzo-15-crown-5, 99%, Thermo Scientific Chemicals

Product Code. 11413337
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11413337

Brand: Thermo Scientific Alfa Aesar A16225.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

Solubility
It is soluble in non-polar solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60835-69-0
Molecular Formula C14H19NO7
Molecular Weight (g/mol) 313.306
MDL Number MFCD00060713
InChI Key XIWRBQVYCZCEPG-UHFFFAOYSA-N
Synonym 4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether
PubChem CID 143747
IUPAC Name 17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene
SMILES C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1

Specifications

Melting Point 93°C to 95°C
Quantity 5 g
Solubility Information It is soluble in non-polar solvents.
Formula Weight 313.3
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzo-15-crown-5

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242017 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog NumberA16225.06,A16225.03
Lot Number10146665 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16225.06,A16225.03
Lot Number10169937 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16225.06,A16225.03
Lot Number10191358 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16225.06,A16225.03
Lot NumberE2213B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16225.06,A16225.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrobenzo-15-crown-5, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.