Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ASTM Certified, Micrometer Pore Sizes
Specifications
Diameter (Metric) Outer | 400 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10146121
|
RETSCH
60.167.000106 |
106μm |
N/A
|
N/A | |||||
10015061
|
RETSCH
60.167.000125 |
125μm |
N/A
|
N/A | |||||
10426621
|
RETSCH
60.167.000180 |
180μm |
N/A
|
N/A | |||||
10166261
|
RETSCH
60.167.000212 |
212μm |
N/A
|
N/A | |||||
10446041
|
RETSCH
60.167.000150 |
150μm |
N/A
|
N/A | |||||
10390282
|
RETSCH
60.167.000250 |
250μm |
N/A
|
N/A | |||||
10176261
|
RETSCH
60.167.000300 |
300μm |
N/A
|
N/A | |||||
10698751
|
RETSCH
60.167.000025 |
25μm |
N/A
|
N/A | |||||
10503342
|
RETSCH
60.167.000032 |
32μm |
N/A
|
N/A | |||||
10330432
|
RETSCH
60.167.000425 |
425μm |
N/A
|
N/A | |||||
10438151
|
RETSCH
60.167.000038 |
38μm |
N/A
|
N/A | |||||
10146361
|
RETSCH
60.167.000355 |
355μm |
N/A
|
N/A | |||||
10310862
|
RETSCH
60.167.000045 |
45μm |
N/A
|
N/A | |||||
10700663
|
RETSCH
60.167.000053 |
53μm |
N/A
|
N/A | |||||
10487381
|
RETSCH
60.167.000500 |
500μm |
N/A
|
N/A | |||||
10640262
|
RETSCH
60.167.000600 |
600μm |
N/A
|
N/A | |||||
10478341
|
RETSCH
60.167.000063 |
63μm |
N/A
|
N/A | |||||
10054941
|
RETSCH
60.167.000710 |
710μm |
N/A
|
N/A | |||||
10418921
|
RETSCH
60.167.000075 |
75μm |
N/A
|
N/A | |||||
10064941
|
RETSCH
60.167.000850 |
850μm |
N/A
|
N/A | |||||
10741043
|
RETSCH
60.167.000090 |
90μm |
N/A
|
N/A | |||||
Specifications
400 mm | |
Stainless Steel | |
1Each | |
ASTM E 11 | |
Shaker AS 450 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
65mm |