missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Pyridyl)-1H-tetrazole, 98%, Thermo Scientific Chemicals

CAS: 33893-89-9 | C6H4N5 | 146.13 g/mol

408.00€

Chemical Identifiers

CAS 33893-89-9
Molecular Formula C6H4N5
Molecular Weight (g/mol) 146.13
MDL Number MFCD00068114
InChI Key ZKHJXYLAESJIEI-UHFFFAOYSA-N
Synonym 5-2-pyridyl-1h-tetrazole, 2-1h-tetrazol-5-yl pyridine, 2-2h-tetrazol-5-yl pyridine, 2-2h-tetrazol-5-yl-pyridine, pyridine, 2-1h-tetrazol-5-yl, 5-2-pyridinyl tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl pyridine, 5-2-pyridyl tetrazole, 5-pyridin-2-yl tetrazole, chembl80094
PubChem CID 320267
SMILES [N-]1N=NN=C1C1=CC=CC=N1
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495796
SDS View Documents
Thermo Scientific Alfa Aesar
B20852.06
5 g
408.00€
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,complex .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,σ-complex .

Solubility
Soluble in water at 25°C 11 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(2-Pyridyl)-1H-tetrazole, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.