missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ 5-(3-Fluorophenyl)-1H-tetrazole, 97% Product Code.: 11378326

Alfa Aesar™ 5-(3-Fluorophenyl)-1H-tetrazole, 97%

Product Code. 11378326
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11378326

Brand: Alfa Aesar™ L15558.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50907-20-5
Molecular Formula C7H5FN4
Molecular Weight (g/mol) 164.143
MDL Number MFCD01075702
InChI Key SEYSTLGMSOLFBD-UHFFFAOYSA-N
Synonym 5-3-fluorophenyl-1h-tetrazole, 5-3-fluorophenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetrazole, 5-3-fluorophenyl-2h-1,2,3,4-tetrazole, 5-3-fluoro-phenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetraazole, acmc-20am9c, 3-fluorophenyl-tetrazole, 5-3-fluorophenyl tetrazoleShow MoreShow Less
PubChem CID 333943
IUPAC Name 5-(3-fluorophenyl)-2H-tetrazole
SMILES C1=CC(=CC(=C1)F)C2=NNN=N2

Specifications

Melting Point 142°C to 144°C
Quantity 250 mg
Beilstein 1104440
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 5-(3-Fluorophenyl)-1H-tetrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 5-(3-Fluorophenyl)-1H-tetrazole, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.