missing translation for 'onlineSavingsMsg'
Learn More
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific Chemicals Product Code.: 11382339

5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific Chemicals

Product Code. 11382339
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11382339

Brand: Thermo Scientific Alfa Aesar H27287.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51516-68-8
Molecular Formula C10H7ClN4
Molecular Weight (g/mol) 218.64
MDL Number MFCD00128298
InChI Key VRKRSWGTAMOYEV-UHFFFAOYSA-N
Synonym 5-amino-1-3-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl-1h-pyrazole-, 5-amino-1-3-chloro-phenyl-1h-pyrazole-4-carbonit, 1h-pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl, 1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl
PubChem CID 734496
IUPAC Name 5-amino-1-(3-chlorophenyl)pyrazole-4-carbonitrile
SMILES NC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N

Specifications

Melting Point 183°C to 187°C
Quantity 1 g
Formula Weight 218.65
Percent Purity 97%
Chemical Name or Material 5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10216105 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberH27287.06,H27287.03
Lot Number10136497 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27287.06,H27287.03
Lot Number10216105 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27287.06,H27287.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.