missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(N-methyl-N-benzylamino)pyridine, 95%, Thermo Scientific™

Product Code. 15428245
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187386-36-2
Molecular Formula C13H13BrN2
Molecular Weight (g/mol) 277.165
MDL Number MFCD12913988
InChI Key WVDSURJOVZYTTF-UHFFFAOYSA-N
Synonym 2-n-benzyl-n-methylamino-5-bromopyridine, n-benzyl-5-bromo-n-methyl-2-pyridinamine, 5-bromo-2-n-methyl-n-benzylamino pyridine, acmc-2099zr
PubChem CID 46739785
IUPAC Name N-benzyl-5-bromo-N-methylpyridin-2-amine
SMILES CN(CC1=CC=CC=C1)C2=NC=C(C=C2)Br

Specifications

Quantity 250 mg
Formula Weight 263.14
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-(N-methyl-N-benzylamino)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'