missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals Product Code.: 15480688

5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals

Product Code. 15480688
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15480688

Brand: Thermo Scientific Alfa Aesar H54386.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10522-47-1
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.08
MDL Number MFCD04966996
InChI Key ZRQISUREPLHYIG-UHFFFAOYSA-N
Synonym quinoline, 5-bromo-8-methoxy
PubChem CID 4715023
IUPAC Name 5-bromo-8-methoxyquinoline
SMILES COC1=C2N=CC=CC2=C(Br)C=C1

Specifications

Melting Point 80°C to 82°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 238.08
Percent Purity 96%
Chemical Name or Material 5-Bromo-8-methoxyquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.