missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | 173°C to 175°C |
Quantity | 5 g |
Solubility Information | Soluble in methanol. |
Formula Weight | 202.01 |
Percent Purity | 98% |
Chemical Name or Material | 5-Bromopyridine-2-carboxylic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Bromopyridine-2-carboxylic acid, 98% >
Spot an opportunity for improvement?Share a Content Correction