missing translation for 'onlineSavingsMsg'
Learn More
5-Fluoro-2-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™ Product Code.: 15495737

5-Fluoro-2-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15495737
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15495737

Brand: Thermo Scientific Alfa Aesar H32956.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an organic fluorinated building block. It participates in the synthesis of diphenylthioethers. It also participates in the preparation of 1-cyano derivative of nilutamide.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092460-81-5
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD11519331
InChI Key DLJQIFHQYJCQHB-UHFFFAOYSA-N
Synonym 5-fluoro-2-trifluoromethoxy benzaldehyde, 5-fluoro-2-trifluoromethoxy benzaldehyde, jrd
PubChem CID 45789803
IUPAC Name 5-fluoro-2-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)OC(F)(F)F

Specifications

Refractive Index 1.4405
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10158123 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10183170 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10164295 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Fluoro-2-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.