missing translation for 'onlineSavingsMsg'
Learn More
5-Methoxy-2-nitrobenzoic acid, 97% Product Code.: 10285431

5-Methoxy-2-nitrobenzoic acid, 97%

Product Code. 10285431 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle, 25g
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10285431

Brand: Thermo Scientific Acros 334930250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1882-69-5 | C8H6NO5 | 196.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1882-69-5
Molecular Formula C8H6NO5
Molecular Weight (g/mol) 196.14
MDL Number MFCD00151836
InChI Key URADKXVAIGMTEG-UHFFFAOYSA-M
Synonym 5-methoxy-2-nitrobenzoic acid, 2-nitro-5-methoxybenzoic acid, 6-nitro-m-anisic acid, 5-methoxy-2-nitro-benzoic acid, benzoic acid, 5-methoxy-2-nitro, pubchem4229, acmc-1brd6, 2-nitro-5-methoxybenzoate, 2-nitro-5-methoxybenzoicacid, 5-methoxy-2-nitrobenzoicacidShow MoreShow Less
PubChem CID 339209
IUPAC Name 5-methoxy-2-nitrobenzoate
SMILES COC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 125°C to 130°C
Color Beige to Gray or Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 197.15
Percent Purity 97%
Physical Form Granules
Chemical Name or Material 5-Methoxy-2-nitrobenzoic acid, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0174752 Certificate TypeCertificate of Analysis Date11/03/2024 Product Code334930250,334930050
Lot NumberA0206675 Certificate TypeCertificate of Analysis Date15/12/2022 Product Code334930250,334930050
compliance-icons
Product Identifier
  • 5-Methoxy-2-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methoxy-2-nitrobenzoic acid, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.