missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[5-(Morpholinomethyl)isoxazol-3-yl]methylamine, 97%, Thermo Scientific™
168.00€ - 612.00€
Chemical Identifiers
CAS | 893748-66-8 |
---|---|
Molecular Formula | C9H15N3O2 |
Molecular Weight (g/mol) | 197.24 |
MDL Number | MFCD06804185 |
InChI Key | JJKIQRMZVVZXSK-UHFFFAOYSA-N |
Synonym | 5-morpholinomethyl isoxazol-3-yl methylamine, 3-isoxazolemethanamine, 5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine, 1-5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamineShow More |
PubChem CID | 20099025 |
IUPAC Name | [5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine |
SMILES | C1COCCN1CC2=CC(=NO2)CN |
Description
Chemical Identifiers
893748-66-8 | |
197.24 | |
JJKIQRMZVVZXSK-UHFFFAOYSA-N | |
20099025 | |
C1COCCN1CC2=CC(=NO2)CN |
C9H15N3O2 | |
MFCD06804185 | |
5-morpholinomethyl isoxazol-3-yl methylamine, 3-isoxazolemethanamine, 5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine, 1-5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamineShow More | |
[5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine |
Specifications
893748-66-8 | |
100.0 | |
Authentic | |
C9H15N3O2 | |
5-morpholinomethyl isoxazol-3-yl methylamine, 3-isoxazolemethanamine, 5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine, 1-5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine | |
C1COCCN1CC2=CC(=NO2)CN | |
197.24 | |
197.24 | |
Liquid |
96.0 | |
Yellow | |
96% min. (HPLC) | |
MFCD06804185 | |
JJKIQRMZVVZXSK-UHFFFAOYSA-N | |
[5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine | |
20099025 | |
97% | |
[5-(Morpholinomethyl)isoxazol-3-yl]methylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- ¢5-(Morpholinomethyl)isoxazol-3-yl!methylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
[5-(Morpholinomethyl)isoxazol-3-yl]methylamine, 97%, Thermo Scientific™