Learn More
5-Phenyl-1-pentyne, 98+%, Thermo Scientific Chemicals Product Code.: 11392517
5-Phenyl-1-pentyne, 98+%, Thermo Scientific Chemicals
Product Code. 11392517
5 g
Click to view available options
Quantity:
5 g
25 g
Product Code. 11392517
Brand: Thermo Scientific Alfa Aesar L11608.06
Request Bulk or Custom Format
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1823-14-9
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD00039813
InChI Key KOSORCNALVBYBP-UHFFFAOYSA-N
Synonym 5-phenyl-1-pentyne, pent-4-yn-1-ylbenzene, benzene, 4-pentynyl, 5-phenylpentyne, unii-b4qv76z86q, pent-4-ynyl-benzene, ccris 7101, pent-4-yn-1-yl benzene, 5-phenyl pentyne, 4-pentynylbenzene #
PubChem CID 74573
IUPAC Name pent-4-ynylbenzene
SMILES C#CCCCC1=CC=CC=C1
Specifications
Density 0.95
Boiling Point 94°C (50 mmHg)
Flash Point 85°C (185°F)
Linear Formula C6H5(CH2)3C≡CH
Refractive Index 1.5165
Quantity 5 g
Beilstein 2074729
Formula Weight 144.22
Percent Purity ≥98%
Chemical Name or Material 5-Phenyl-1-pentyne
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW24K042 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberL11608.06,L11608.14
Lot Number10252122 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog NumberL11608.06,L11608.14
Lot NumberX29H048 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL11608.06,L11608.14
Lot Number10245943 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL11608.06,L11608.14
Lot NumberZ06I034 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog NumberL11608.06,L11608.14
Product Title
5-Phenyl-1-pentyne, 98+%, Thermo Scientific Chemicals >