Learn More
Agilent BioTek 50 TS Microplate Washer
Alle ansehen Agilent BioTek Produkte
Beschreibung
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Garantie
12 months
Spezifikation
Spezifikation
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Connections | USB (1) |
| Bottles | Optional (2 to 4 L) |
| Plate Type | 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) |
| Soak Time | Programmable in minutes and seconds, up to 30 min. |
| Type | Microplate Washer |
| Weight (Metric) | 9.8 kg |
| Wash Cycles | 1 to 10 |
| Wash Volume | 25 to 3000 μL/well |
| Display | 4.3 in. LCD Color Touch Screen |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.