Learn More
Fisher Chemical 500ml Buffer Solution, Colour Coded Red, pH 4 to NIST, 500ml
Brand: Fisher Chemical J/2826/08
23.14 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
500ml Buffer Solution, Colour Coded Red, pH 4 to NIST, 500ml
Description
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30