Learn More
Univet™ 506UP Hybrid Clear Safety Glasses
506 UP Hybrid has lenses come complete with a highly effective anti-fog and anti-scratch coating. Adjustable temples and soft rubber seal ensure a smooth fit on the face and nose.
Brand: Univet™ 506UG.03.00.00
This item is not returnable.
View return policy
Description
- Lens with ultra performing anti-fog and anti-scratch Vanguard PLUS treatment
- Temples adjustable in inclination
- Soft-rubber gasket for a gentle support on the face
- Panoramic and wraparound lens for a better side protection
- Adjustable elastic headband provides additional protection
Specifications
Rectangular | |
None | |
Polycarbonate | |
Polycarbonate | |
UV380 | |
Not Autoclavable |
Anti-fog, Scratch-resistant | |
Safety Glasses | |
Reusable | |
None | |
Adjustable Temples |