missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 6-Bromo-2(3H)-benzoxazolone, 97%
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 193°C to 195°C |
Quantity | 1 g |
Formula Weight | 214 |
Percent Purity | 97% |
Chemical Name or Material | 6-Bromo-2(3H)-benzoxazolone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'