missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1-indanone, 97%, Thermo Scientific™

Product Code. 11954861
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1481-32-9
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.15
MDL Number MFCD01318147
InChI Key LVUUCFIQQHEFEJ-UHFFFAOYSA-N
Synonym 6-fluoro-1-indanone, 6-fluoro-2,3-dihydro-1h-inden-1-one, 6-fluoro-indan-1-one, 6-fluoroindan-1-one, 6-fluroro-1-indanone, 1h-inden-1-one, 6-fluoro-2,3-dihydro, 6-fluoroindanone, 6fluoro-1-indanone, pubchem8856, acmc-1bwfy
PubChem CID 1519464
IUPAC Name 6-fluoro-2,3-dihydroinden-1-one
SMILES FC1=CC2=C(CCC2=O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 61°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 150.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-1-indanone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Fluoro-1-indanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Fluoro-1-indanone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.