missing translation for 'onlineSavingsMsg'
Learn More
6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals Product Code.: 11332766

6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals

Product Code. 11332766
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11332766

Brand: Thermo Scientific Alfa Aesar L08877.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3222-47-7 | C7H7NO2 | 137.138 g/mol

6-Methylnicotinic acid is an intermediate of the drug etoricoxib (a non- steroidal anti-inflammatory drug for the treatment of arthritis and osteoarthritis). It is also used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylnicotinic acid is an intermediate of the drug etoricoxib (a non- steroidal anti-inflammatory drug for the treatment of arthritis and osteoarthritis). It is also used as organic intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3222-47-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00006341
InChI Key RZOKQIPOABEQAM-UHFFFAOYSA-N
Synonym 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218
PubChem CID 137860
IUPAC Name 6-methylpyridine-3-carboxylic acid
SMILES CC1=NC=C(C=C1)C(=O)O

Specifications

Melting Point 210°C to 215°C
Quantity 1 g
Beilstein 112049
Solubility Information Sparingly soluble in water.
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material 6-Methylnicotinic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10167798 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL08877.06,L08877.03,L08877.14
Lot NumberM02K025 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberL08877.06,L08877.03,L08877.14
Lot NumberT13I010 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog NumberL08877.06,L08877.03,L08877.14
Lot NumberGBFY034213 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL08877.06,L08877.03,L08877.14
Lot Number10156218 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL08877.06,L08877.03,L08877.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.